Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563412
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563412 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563412 Supplier Invitrogen™ Supplier No. PA563412
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CCPLLEQPNI VHDLPAAVLS YCQVWKIPAI LYLCYTDVMK LDLITVEAFK PILSTRSLKG LVKNIPQSTE ILKKLMTTNE IQSNIYT Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 87%.

Chaperone protein which promotes assembly of the 20S proteasome as part of a heterodimer with PSMG2. The PSMG1-PSMG2 heterodimer binds to the PSMA5 and PSMA7 proteasome subunits, promotes assembly of the proteasome alpha subunits into the heteroheptameric alpha ring and prevents alpha ring dimerization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMG1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PSMG1
Gene Accession No. O95456
Gene Alias AW552102; C21LRP; C21-LRP; Chromosome 21 leucine-rich protein; Down syndrome critical region gene 2; Down syndrome critical region homolog 2; Down syndrome critical region protein 2; Down syndrome critical region protein 2 homolog; Dscr2; leucine rich protein C21-LRP; LRPC21; PAC1; PAC-1; proteasome (prosome, macropain) assembly chaperone 1; proteasome assembling chaperone 1; proteasome assembly chaperone 1; Psmg1
Gene Symbols PSMG1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PSMG1. Recombinant protein control fragment (Product #RP-91798).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8624
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.