Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ PSMG4 Protein
SDP

Catalog No. NBP17881310 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17881310 100μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP17881310 Supplier Novus Biologicals™ Supplier No. NBP178813100UG
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-123 of Human PSMG4 The Recombinant Human PSMG4 Protein is derived from E. coli. The Recombinant Human PSMG4 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.5mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 10% glycerol, 0.1M NaCl
Molecular Weight (g/mol) 15.9kDa
Purification Method SDS-PAGE
Quantity 100μg
Source Human
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name PSMG4
Conjugate Unconjugated
Purity or Quality Grade >95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.