Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSRC1 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501735571 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5571 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5571 Supplier Proteintech Group Inc Supplier No. 271301AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a proline-rich protein. Studies of this gene and the related mouse gene suggest that this gene is regulated by p53 and may participate in p53-mediated growth suppression. The encoded protein may function as a microtubule destabilizing protein that controls spindle dynamics and mitotic progression by recruiting and regulating microtubule depolymerases. At least one genetic variation in this gene has been associated with decreased serum levels of low-density lipoprotein cholesterol. Alternatively spliced transcript variants encoding different isoforms have been described.

Specifications

Antigen PSRC1
Applications Western Blot
Classification Polyclonal
Concentration 0.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PSRC1
Gene Accession No. Q6PGN9
Gene Alias DDA3, FP3214, PSRC1, RP11 297O4.2
Gene Symbols PSRC1
Host Species Rabbit
Immunogen PSRC1 Fusion Protein Ag26039
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84722
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.