Learn More
Selleck Chemical LLC PTC-209 2mg 315704-66-6
SDP
Supplier:  Selleck Chemical LLC S73722MG
Encompass_Preferred
PTC-209 is a potent and selective BMI-1 inhibitor with IC50 of 0.5 ?M in HEK293T cell line, and results in irreversible reduction of cancer-initiating cells (CICs). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8026
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.