Learn More
Apexbio Technology LLC PTC-209 315704-66-6 50mg
SDP
Supplier:  Apexbio Technology LLC B317950
Encompass_Preferred
PTC-209 (CAS 315704-66-6) is a small-molecule inhibitor targeting BMI-1 identified through Gene Expression Modulation by Small molecules (GEMS) technology It acts by suppressing BMI-1 expression at the transcriptional level displaying an IC50 of approximately 0 5 M In HCT116 cells PTC-209 reduces both UTR-mediated and endogenous BMI-1 expression It selectively restricts cell proliferation in certain tumor and stem cell types including U937 HT1080 and human hematopoietic stem cells without affecting viability in cell lines such as HEK293 and HT1080 Thus PTC-209 is primarily utilized in studies exploring cancer stem cell growth and oncogenesis involving BMI-1 and the PRC1 complex
Catalog No. 50-000-62798
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.