Learn More
World Precision Instrument Patch Clamp Glass-PG52151-4
SDP
Supplier:  World Precision Instrument PG521514
Encompass
PG52151-4 and PG52165-4 are prepared from Schott #8250 glass (equivalent to Corn­ing #7052), one of the most widely used patch clamp­ing glasses. This is a specially formulated boro­sil­i­cate glass with a softening temperature that is 110°C lower than regular borosilicate glass (Corn­ing 7740, or Pyrex). It has excellent sealing prop­er­ties for most cells. Electrical properties are also very good.
Catalog No. 50-821-929