Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTER Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558226
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558226 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558226 Supplier Invitrogen™ Supplier No. PA558226
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DATHSSETRA MSVEQLTDVL MNEILHGADG TSIKCGIIGE IGCSWPLTES ERKVLQATAH AQAQLGCPVI I Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 92%.

PTER is a mammalian homolog to bacterial phosphotriesterases, enzymes that hydrolyze phosphotriester-containing organophosphate pesticides. It is expressed primarily in the proximal renal tubules and the gene has been localized in humans to chromosomal band 10p12 by in situ hybridization. PTER, in addition to FTO, MC4R, and NPC1 has recently been shown to be a new risk loci for early-onset and morbid adult obesity in European populations. At least two isoforms of PTER are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTER
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PTER
Gene Accession No. Q96BW5
Gene Alias AI790318; HPHRP; Mpr56-1; Parathion hydrolase-related protein; phosphotriesterase related; phosphotriesterase-related protein; Pter; resiniferatoxin-binding phosphotriesterase-related protein; resiniferatoxin-binding, phosphotriesterase-related; resiniferatoxin-binding, phosphotriesterase-related protein; resiniferotoxin-binding phosphotriesterase-related protein; Rpr1; Rpr-1
Gene Symbols PTER
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PTER. Recombinant protein control fragment (Product #RP-97145).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9317
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.