Learn More
Enzo Life Sciences Pterostilbene (100mg). CAS: 537-42-8
SDP
Supplier:  Enzo Life Sciences ALX385034M100
Encompass_Preferred
Cell permeable natural methoxylated analog of resveratrol (BML-FR104). Antioxidant, antiproliferative, anti-inflammatory, anti-hyperglycemic and anti-diabetic agent. Induces apoptosis. Induces Autophagy. Very potent inhibitor of cytochrome P450 1A1 (CYP1A1) with Ki=0.57µM.Inhibits activator protein 1 (AP-1) and NF-κB activation. Moderately inhibits COX-1 and COX-2 (IC50=19.8µM and 83.9µM). Alternative name: trans-3,5-Dimethoxy-4'-hydroxystilbene. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (20mg/ml) or methanol; insoluble in water. Long Term Storage: +4°C.
Catalog No. 50-200-9062
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.