Learn More
TARGETMOL CHEMICALS INC PTERYXIN 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T5S140810MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (+)-Pteryxin ((+)-Pteryxin) has muscle-relaxant props. (+)-Pteryxin shows hepatoprotective and nitric oxide prodn. inhibitory activity. purity: 99%
Catalog No. 50-257-6639