Learn More
BRANDTECH™ BRAND™ PTFE Closure Ring for Dispensette™ Bottletop Dispensers
Catalog No. 13880724 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Material:
PTFE
Catalog No. Material
13-880-724 PTFE
1 options
Catalog No. 13-880-724 Supplier BRANDTECH™ BRAND™ Supplier No. 704486
Add to Cart
Add to Cart
For Sensitive chemicals
  • PTFE ring used to seal rotating valve block on Dispensette™ dispensers made after 1997
TRUSTED_SUSTAINABILITY
Specifications
Description For use with highly volatile reagents
Material PTFE
For Use With (Equipment) Dispensette Valve Block
WARNING: Reproductive Harm - www.P65Warnings.ca.gov