Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ PTFE Disposable Stir Bars PROMO

Catalog No. 1451393PM Shop All Fisherbrand Products
Change view
Click to view available options
Length (English):
0.5 in.
1 in.
1.61 in.
2 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
14-513-93PM 0.5 in.
14-513-94PM 1 in.
14-513-95PM 1.61 in.
14-513-83PM 2 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-513-93PM

The multifaceted magnetic stirring bar surface adds turbulence relative to similar size smooth cylindrical stirring bars.

  • Ideal for single use applications mixing radioactive reagents or when cross contamination might be a concern.
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE low friction coating; durable and inert.
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.

Specifications

Color White
Type Stir Bar
Coating PTFE
Length (Metric) 12.7 mm
Length (English) 0.5 in.
Diameter (English) 0.125 in.
Diameter (Metric) 3.2 mm
Certifications/Compliance ISO 9001:2008
Material PTFE
Quantity 100
Autoclavable Autoclavable
Shape Cylinder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.