Learn More
Cytiva (Formerly Pall Lab) PTFE Membrane Disc Filters - 2 m, 47 mm (50/pkg)
Catalog No. 502063493 Shop All Cytiva Products
Encompass
Strong, Chemically Resistant Membranes for Air Monitoring and Sampling in Aggressive EnvironmentsLow chemical background permits highly sensitive, interference-free determinations.Ensures accurate gravimetric determinations with low tare mass.Ideal for filtration of gas and/or organic solvents. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3493 Supplier Cytiva Supplier No. R2PJ047
Strong, Chemically Resistant Membranes for Air Monitoring and Sampling in Aggressive EnvironmentsLow chemical background permits highly sensitive, interference-free determinations.Ensures accurate gravimetric determinations with low tare mass.Ideal for filtration of gas and/or organic solvents. This product is not approved for use in medical, clinical, surgical or other patient applications.