Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ PTFE Packed GC Column (General Configuration), Phase: None, Matrix Chromosil 330 Support

Catalog No. 111009168 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
2.4 m
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-100-9168 2.4 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-9168 Supplier MilliporeSigma™ Supelco™ Supplier No. 11496
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For low temperature applications, a PTFE column offers the flexibility of metal with inertness approaching that of glass. This column is of a general configuration, and can be carefully bent to fit most instruments.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Gas Chromatography (GC)
Length (Metric) 2.4 m
Material PTFE
Packing Material Matrix Chromosil 330 Support
Diameter (Metric) Inner 2.1 mm
Diameter (English) Outer 0.125 in.
Product Type Packed GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.