Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTGER2/PTGER3 Monoclonal Antibody (3F12)
GREENER_CHOICE

Catalog No. 01674033 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-033 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-033 Supplier Invitrogen™ Supplier No. MA112677
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

A suggested positive control for this product is porcine brain lysate.

This polyclonal antibody was raised in Rabbit against PTGER3 / EP3. This antibody has been conjugated with Unconjugated. This antibody is useful for testing in the following assays: IHC-P. The specificity of this antibody has been described as follows: Human PTGER3 / EP3. BLAST analysis of the peptide immunogen showed no homology with other human proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTGER2/PTGER3
Applications Western Blot
Classification Monoclonal
Clone 3F12
Concentration 0.11 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene Ptger3
Gene Accession No. P30557, P34980, P43115, P43116, Q62053, Q62928
Gene Alias EP2; EP2 receptor; EP3; EP3e; EP3-I; EP3-II; EP3-III; EP3-IV; EP3-VI; PGE receptor EP2 subtype; PGE receptor EP3 subtype; PGE receptor, EP2 subtype; PGE receptor, EP3 subtype; PGE2 receptor EP2 subtype; PGE2 receptor EP3 subtype; PGE2-R; Pgerep3; prostaglandin E receotor EP3 subtype 3 isoform; prostaglandin E receptor 2; prostaglandin E receptor 2 (EP2 subtype); prostaglandin E receptor 2 (subtype EP2); prostaglandin E receptor 2 (subtype EP2), 53kD; prostaglandin E receptor 2 (subtype EP2), 53kDa; prostaglandin E receptor 3; prostaglandin E receptor 3 (subtype EP3); prostaglandin E receptor EP2 subtype; prostaglandin E receptor EP3 subtype; prostaglandin E receptor subtype EP2; prostaglandin E2 receptor EP2 subtype; Prostaglandin E2 receptor EP3 subtype; prostaglandin E2 receptor type 2; prostaglandin receptor (PGE-2); prostanoid EP2 receptor; Prostanoid EP3 receptor; PTGER2; Ptger3; Ptgerep2; Ptger-ep2; Ptgerep3; Rep3; rEP3a; rEP3b
Gene Symbols Ptger2, Ptger3
Host Species Mouse
Immunogen Recombinant human EP2 and EP3 receptor protein.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19217, 19218, 24929, 5732, 5733, 81752
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.