Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTGER3 Monoclonal Antibody (5F5)
GREENER_CHOICE

Catalog No. 01674028
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-028 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-028 Supplier Invitrogen™ Supplier No. MA112647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

A suggested positive control for this product is porcine brain lysate.

CD172a, also known as signal-regulatory protein alpha, is a receptor-type transmembrane glycoprotein expressed on cells of myeloid origin, including granulocytes, dendritic cells (DCs), macrophages, mast cells and haematopoietic stem cells. CD172a acts as a substrate for several activated tyrosine kinases, including EGFR, PDGFR, src and insulin receptor and is involved in the negative regulation of receptor tyrosine kinase-coupled signaling pathways. Ligand binding of CD172a to integrin-associated protein CD47, results in tyrosine kinase phosphorylation of immunoreceptor tyrosine-based inhibitory motifs (ITIMs) within the cytoplasmic region of CD172a, mediating the recruitment and activation of the tyrosine phosphatases SHP-1 and SHP-2. These then act as regulators of cellular function, through dephosphorylation of specific substrates. Ligation of CD172a with CD47 has been demonstrated in several regulatory processes, including the inhibition of host cell phagocytosis by macrophages and the bi-directional activation of T cells and DCs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTGER3
Applications Western Blot
Classification Monoclonal
Clone 5F5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene PTGER3
Gene Accession No. P34979, P34980, P43115
Gene Alias EP3; Ep3 receptor; EP3A receptor; EP3B receptor; EP3C receptor; EP3D receptor; EP3e; EP3-I; EP3-II; EP3-III; EP3-IV; EP3-VI; MGC141828; MGC141829; MGC27302; PGE receptor EP3 subtype; PGE receptor, EP3 subtype; PGE2 receptor EP3 subtype; PGE2-R; Pgerep3; pr; prostaglandin E receotor EP3 subtype 3 isoform; prostaglandin E receptor 3; prostaglandin E receptor 3 (subtype EP3); prostaglandin E receptor EP3 subtype; prostaglandin E2 receptor EP3 subtype; prostaglandin receptor (PGE-2); prostanoid EP3 receptor; PTGER3; Ptgerep3; Rep3; rEP3a; rEP3b
Gene Symbols PTGER3
Host Species Mouse
Immunogen Recombinant human EP3 Receptor.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24929, 282330, 5733
Target Species Human, Rat, Bovine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.