Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTGER3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. p-7211020 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4527 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4527 Supplier Proteintech Group Inc Supplier No. 247611AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Receptor for prostaglandin E2 (PGE2); the EP3 receptor may be involved in inhibition of gastric acid secretion, modulation of neurotransmitter release in central and peripheral neurons, inhibition of sodium and water reabsorption in kidney tubulus and contraction in uterine smooth muscle. The activity of this receptor can couple to both the inhibition of adenylate cyclase mediated by G-I proteins, and to an elevation of intracellular calcium. The various isoforms have identical ligand binding properties but can interact with different second messenger systems.

Specifications

Antigen PTGER3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PTGER3
Gene Accession No. P30557, P43115
Gene Alias EP3, EP3 I, EP3 II, EP3 III, EP3 IV, EP3e, PGE receptor EP3 subtype, PGE2 R, PGE2 receptor EP3 subtype, Prostanoid EP3 receptor, PTGER3
Gene Symbols PTGER3
Host Species Rabbit
Immunogen PTGER3 Fusion Protein Ag21542
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19218, 5733
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.