Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTH Monoclonal Antibody (BGN/1F8)
GREENER_CHOICE

Catalog No. PIMA183385
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA183385 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA183385 Supplier Invitrogen™ Supplier No. MA183385
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Mouse anti Human parathyroid hormone monoclonal antibody, clone BGN/1F8 recognizes human parathyroid hormone (PTH) also known as Parathyrin.

This gene encodes a member of the parathyroid family of proteins. The encoded preproprotein is proteolytically processed to generate a protein that binds to the parathyroid hormone/parathyroid hormone-related peptide receptor and regulates blood calcium and phosphate levels. Excess production of the encoded protein, known as hyperparathyroidism, can result in hypercalcemia and kidney stones. On the other hand, defective processing of the encoded protein may lead to hypoparathyroidism, which can result in hypocalcemia and numbness.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTH
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone BGN/1F8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene PTH
Gene Accession No. P01270, P04089
Gene Alias hypothalamic parathyroid hormone; parathormone; Parathyrin; parathyroid hormone; parathyroid hormone 1; preproparathyroid hormone; prepro-PTH; PTH; PTH-(1-84); Pth1; Pthp; Pthr1
Gene Symbols PTH
Host Species Mouse
Immunogen Synthetic peptide corresponding to amino acids 1-34 of mature PTH conjugated to a a proprietary carrier molecule.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19226, 24694, 5741
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.