Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTH Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. p-7208334 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-2316 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-2316 Supplier Proteintech Group Inc Supplier No. 190011AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PTH is a hormone producted by the parathyroid gland that regulates the concentration of calcium and phosphorus in extracellular fluid. It is produced in the parathyroid gland as an 84 amino acid single chain polypeptide. Defects in this gene are a cause of familial isolated hypoparathyroidism.

Specifications

Antigen PTH
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PTH
Gene Accession No. P01270
Gene Alias Parathormone, Parathyrin, parathyroid hormone, PTH, PTH1
Gene Symbols PTH
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5741
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.