Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PeproTech PTHLH Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Catalog No. 500P276BT25
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μg
50 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
500P276BT25 25 μg
500P276BT50 50 μg
50-271-8545 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 500P276BT25 Supplier PeproTech Supplier No. 500P276BT25UG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: AVSEHQLLHD KGKSIQDLRR RFFLHHLIAE IHTAEIRATS EVSPNSKPSP NTKNHPVRFG SDDEGRYLTQ ETNKVETYKE QPLKTP. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human PTHrP. Anti-Human PTHrP-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hPTHrP by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human PTHrP (500-P276) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hPTHrP. Western Blot: To detect hPTHrP by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hPTHrP is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P276BT-1MG will be provided as 2 x 500 μg

The protein encoded by this gene is a member of the parathyroid hormone family. This hormone regulates endochondral bone development and epithelial-mesenchymal interactions during the formation of the mammary glands and teeth. This hormone is involved in lactation possibly by regulating the mobilization and transfer of calcium to the milk. The receptor of this hormone, PTHR1, is responsible for most cases of humoral hypercalcemia of malignancy. Four alternatively spliced transcript variants encoding two distinct isoforms have been observed. There is also evidence for alternative translation initiation from non-AUG (CUG and GUG) start sites, in-frame and downstream of the initiator AUG codon, to give rise to nuclear forms of this hormone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTHLH
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Biotin
Formulation PBS with no preservative
Gene PTHLH
Gene Accession No. P12272
Gene Alias BDE2; calciotropic hormone, growth factor; HHM; osteostatin; parathyroid hormone like hormone; parathyroid hormone related peptide; parathyroid hormone-like hormone; parathyroid hormone-like hormone preproprotein; parathyroid hormone-like peptide; parathyroid hormone-like protein; parathyroid hormone-like related protein; parathyroid hormone-related peptide; parathyroid hormone-related protein; parathyroid-like peptide; PLP; PTHLH; PTH-like; PTHR; PTH-related peptide; PTH-related protein; Pthrp; PTH-rP; PTHrP[107-139]; PTHrP[1-36]; PTHrP[38-94]
Gene Symbols PTHLH
Host Species Rabbit
Immunogen E.coli-derived, 9.8 kDa Recombinant Human PTHrP.
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5744
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.