Learn More
Enzo Life Sciences PTIO (100mg). CAS: 18390-00-6
SDP
Supplier:  Enzo Life Sciences ALX430007M100
Encompass_Preferred
Stable nitric oxide (NO) radical scavenger. Alternative name: 2-Phenyl-4,4,5,5-tetramethylimidazoline-1-oxyl-3-oxide. Purity: ≥98% (Nonaqueous Titration). Solubility: Soluble in methanol, 100% ethanol or water. Long Term Storage: -20°C.
Catalog No. 50-200-9132
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.