Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTMA Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01674415
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-415 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-415 Supplier Invitrogen™ Supplier No. PA19532
Only null left
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

PA1-9532 detects Thymosin in human, mouse, pig and rat samples. PA1-9532 has been successfully used in ELISA procedures. PA1-9532 immunogen is a synthetic peptide based on human thymosin protein.

The thymus gland produces several hormones or hormone-like substances which are derived from a polypeptide precursor containing (in the rat) 113 amino acids and known as prothymosin-alpha. A peptide containing 28 amino acid residues, named thymosin-alpha-1, was originally isolated from calf thymosin fraction 5 and shown to restore various aspects of immune function in several in vitro and invivo test systems. Thymosin-alpha-1 was subsequently isolated from a similar fraction from human thymosin and reported to have the same amino acid sequence as bovine thymosin-alpha-1. In 2003, Jiang et al. identified a pathway that regulates mitochondria-initiated caspase activity. In this pathway, putative human HLA class II-associated protein (PHAP) promoted caspase-9 activation after apoptosome formation, whereas prothymosin-alpha-1 negatively regulated caspase-9 activation by inhibiting apoptosome formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTMA
Applications ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene PTMA
Gene Accession No. P06302, P06454, P26350
Gene Alias alpha-prothymosin; gene sequence 28; prothymosin alpha; prothymosin alpha protein; Prothymosin alpha, N-terminally processed; prothymosin, alpha; prothymosin-alpha; PTMA; Thym; thymosin; Thymosin alpha; Thymosin alpha-1; TMSA; TMSB4; TMSB4X
Gene Symbols PTMA
Host Species Chicken
Immunogen Synthetic peptide based on human thymosin protein.
Purification Method Affinity chromatography
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100294684, 19231, 29222, 5757
Target Species Human, Mouse, Rat, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.