Learn More
Sigma Organic Chemistry p-Toluenesulfonic acid monohydrate | 15KG | 6192-52-5 | MFCD00142137
Supplier: Sigma Organic Chemistry T3592015KG
            p-Toluenesulfonic acid monohydrate, 15KG
About This Item:
Linear Formula: CH3C6H4SO3H -+ H2O
Storage: room temp
EINECS Number: 203-180-0
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.