Learn More
Eurofins Discoverx PTP-1B, 10 µg. (14-621)
Supplier:  Eurofins Discoverx 14621
Encompass
The PTP-1B product is purified N terminal GST-tagged recombinant PTP-1B enzyme. This enzyme contains amino acids 1-321. It retains its binding and enzymatic activity and is intended for use in phosphatase assays. This enzyme is provided as 10 μg of 0.63 mg/mL frozen solution. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9069
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.