Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTP zeta/Phosphacan Rabbit anti-Rat, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABN224625UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN2246-25UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN2246-25UL Supplier MilliporeSigma Supplier No. ABN224625UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-phosphacan, Cat. No. ABN2246 is a rabbit polyclonal antibody that has been tested for use in Immunohistochemistry (Paraffin) for the detection of Receptor-type tyrosine-protein phosphatase zeta.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTP zeta/Phosphacan
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Rabbit polyclonal antiserum with 0.02% sodium azide.
Gene Alias Receptor-type tyrosine-protein phosphatase zeta;EC:3.1.3.48;R-PTP-zeta;3F8 chondroitin sulfate proteoglycan;3H1 keratan sulfate proteoglycan;Phosphacan
Gene Symbols Ptprz1;Ptprz;Ptpz
Host Species Rabbit
Immunogen Full length (aa 1-2316) purified Receptor-type tyrosine-protein phosphatase zeta (Phosphacan) isolated from 7-day fetal rat brain.
Molecular Weight of Antigen 255.34 kDa calculated.
Purification Method Unpurified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody specifically detects PTP zeta/Phosphacan in rat brain tissue.
Target Species Rat
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Product Type Antibodies
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.