Learn More
Enzo Life Sciences PTP1B (human), (recombinant) (50 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE3320050
Encompass_Preferred
Alternative name: Protein tyrosine phosphatase 1B. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In 50mM HEPES, pH7.2, containing 1mM EDTA, 5mM DTT, and 0.05% NP-40. Source: Produced in E. coli. Human PTP1B (protein tyrosine phosphatase 1B) (aa 1-322). Long Term Storage: -80°C.
Catalog No. 50-201-1986
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.