Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTP4A2 Mouse anti-Human, Clone:5B6F4, Abnova™

Catalog No. 89334592 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-592 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-592 Supplier Abnova Corporation Supplier No. MAB17779
Only null left
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human PTP4A2.

The protein encoded by this gene belongs to a small class of the protein tyrosine phosphatase (PTP) family. PTPs are cell signaling molecules that play regulatory roles in a variety of cellular processes. PTPs in this class contain a protein tyrosine phosphatase catalytic domain and a characteristic C-terminal prenylation motif. This PTP has been shown to primarily associate with plasmic and endosomal membrane through its C-terminal prenylation. This PTP was found to interact with the beta-subunit of Rab geranylgeranyltransferase II (beta GGT II), and thus may function as a regulator of GGT II activity. Overexpression of this gene in mammalian cells conferred a transformed phenotype, which suggested its role in tumorigenesis. Alternatively spliced transcript variants that encode two distinct isoforms have been described. [provided by RefSeq]

Specifications

Antigen PTP4A2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5B6F4
Conjugate Unconjugated
Description protein tyrosine phosphatase type IVA, member 2
Dilution ELISA (1:10000)
Flow Cytometry
Immunocytochemistry
Immunohistochemistry
Western Blot (1:500-1:2000)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene PTP4A2
Gene Alias HH13, HH7-2, HU-PP-1, OV-1, PRL-2, PRL2, PTP4A, PTPCAAX2, ptp-IV1a, ptp-IV1b
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 58-162 of human PTP4A2 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 8073
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.