Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPN1 Antibody, ProSci
SDP

Catalog No. 16831908
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-831-908 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-831-908 Supplier ProSci LLC Supplier No. 300600100
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PTPN1 Polyclonal Antibody specifically detects PTPN1 in Human, Mouse, Rat samples. It is validated for ELISA, Western Blot

Specifications

Antigen PTPN1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution PTPN1 antibody can be used for detection of PTPN1 by ELISA at 1:62500. PTPN1 antibody can be used for detection of PTPN1 by western blot at 0.5 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Formulation Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
Gene Accession No. NP_002818
Gene Alias PTPN1, PTP1B
Host Species Rabbit
Immunogen Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human PTPN1.
Purification Method Antibody is purified by peptide affinity chromatography method.
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5770
Target Species Human, Mouse, Rat
Content And Storage For short periods of storage (days) store at 4°C. For longer periods of storage, store PTPN1 antibody at -20°C. As with any antibody avoid repeat freeze-thaw cycles.
Form Liquid
Research Category Membrane, Cancer, Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.