Learn More
Invitrogen™ PTPN12 Monoclonal Antibody (AG25)

Description
A suggested positive control for this product is SK-N-SH cell lysate.
Specifications
Specifications
| Antigen | PTPN12 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | AG25 |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Gene | PTPN12 |
| Gene Accession No. | P35831, Q05209 |
| Gene Alias | MPTP-PEST; P19-PTP; pest; protein tyrosine phosphatase non-receptor type 12; protein tyrosine phosphatase, non-receptor type 12; protein-tyrosine phosphatase G1; Protein-tyrosine phosphatase P19; PTPG1; PTPN12; PTP-P19; PTP-PEST; RKPTP; tcag7.1075; Tyrosine-protein phosphatase non-receptor type 12 |
| Gene Symbols | PTPN12 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.