Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTPN23 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA592262
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA592262 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA592262 Supplier Invitrogen™ Supplier No. PA592262
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSNRVVLRSG KDDYINASCV EGLSPYCPPL VATQAPLPGT AADFWLMVHE QKVSVIVMLV SEAEMEKQKV ARYFPTERGQ PMVHGALSLA LSSVRSTETH VERVLSLQFR DQSLKRSLVH LHFPTWPELG LPDSPSNLLR FIQEVHAHYL HQRPLHTPII VHCSSGVGRT GAFALLYAAV QEVEAGNGIP ELPQLVRRMR QQRKHMLQEK LHLRFCYEAV V; Positive Samples: Mouse eye, Rat kidney; Cellular Location: Cytoplasm, Cytoplasmic vesicle, Early endosome, Endosome, Nucleus, cilium basal body, cytoskeleton.

Plays a role in sorting of endocytic ubiquitinated cargos into multivesicular bodies (MVBs) via its interaction with the ESCRT-I complex (endosomal sorting complex required for transport I), and possibly also other ESCRT complexes. May act as a negative regulator of Ras-mediated mitogenic activity. Plays a role in ciliogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPN23
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.51 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PTPN23
Gene Accession No. O88902, Q6PB44
Gene Alias AI462446; HDPTP; HD-PTP; His domain-containing protein tyrosine phosphatase; Kiaa1471; Protein tyrosine phosphatase TD14; protein tyrosine phosphatase, non-receptor type 23; PTPN23; PTP-TD14; Tyrosine-protein phosphatase non-receptor type 23
Gene Symbols PTPN23
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1230-1450 of human PTPN23 (NP_0562811).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104831, 117552
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.