Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ PTPRK Recombinant Protein Antigen
SDP

Catalog No. NBP230977PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP230977PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP230977PE Supplier Novus Biologicals™ Supplier No. NBP230977PEP
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human PTPRK. The PTPRK Recombinant Protein Antigen is derived from E. coli. The PTPRK Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.

This is a blocking peptide for NBP2-30977. This is a human recombinant protein expressed in E. coli with a N-terminal His-ABP tag and purified with IMAC chromatography.

Specifications

Gene ID (Entrez) 5796
Species Human
Purification Method Chromatography
Purity >80%
Concentration 0.5mg/mL
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Formulation PBS and 1M Urea, pH 7.4.
For Use With (Application) Blocking/Neutralizing, Control
Gene Alias DKFZp686C2268, DKFZp779N1045, EC 3.1.3.48, protein tyrosine phosphatase kappa protein tyrosine phosphatase kappa, protein tyrosine phosphatase, receptor type, K, Protein-tyrosine phosphatase kappa, protein-tyrosine phosphatase, receptor type, kappa, PTPK, receptor type, K (R-PTP-KAPPA, receptor-type tyrosine-protein phosphatase kappa
Gene Symbol PTPRK
Label Type Unlabeled
Molecular Weight (g/mol) 24kDa
Product Type PTPRK
Quantity 0.1 mL
Regulatory Status RUO
Source E.Coli
Specific Reactivity This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-30977. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Immunogen MTHMVNAMDRSYADQSTLHAEDPLSITFMDQHNFSPRYENHSATAESSRLLDVPRYLCE
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.