Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTPRM Monoclonal Antibody (1H6D10)
GREENER_CHOICE

Catalog No. PIMA531874
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIMA531874 100 μL
1 options

Catalog No. PIMA531874

Supplier: Invitrogen™ MA531874

Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31874 has been tested in indirect ELISA.

The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP possesses an extracellular region, a single transmembrane region, and two tandem catalytic domains, and thus represents a receptor-type PTP. The extracellular region contains a meprin-A5 antigen-PTP mu (MAM) domain, an Ig-like domain and four fibronectin type III-like repeats. This PTP has been shown to mediate cell-cell aggregation through the interaction with another molecule of this PTP on an adjacent cell. This PTP can interact with scaffolding protein RACK1/GNB2L1, which may be necessary for the downstream signaling in response to cell-cell adhesion. Alternative splicing results in multiple transcripts encoding distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPRM
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1H6D10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene PTPRM
Gene Accession No. P28827
Gene Alias hR-PTPu; Kiaa4044; MGC166994; mKIAA4044; protein tyrosine phosphatase mu; protein tyrosine phosphatase receptor type M; protein tyrosine phosphatase receptor-type M; protein tyrosine phosphatase, receptor type M; protein tyrosine phosphatase, receptor type, M; protein tyrosine phosphatase, receptor type, mu polypeptide; protein tyrosine phosphatase, receptor-type, M; protein-tyrosine phosphatase mu; PTP mu; Ptpmu; PTPRL1; PTPRM; receptor-type tyrosine-protein phosphatase mu; RPTPM; RPTPmu; R-PTP-MU; RPTPU
Gene Symbols PTPRM
Host Species Mouse
Immunogen Purified recombinant fragment of human PTPRM (AA: extra 565-742) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5797
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.