Learn More
Enzo Life Sciences PTX3 monoclonal antibody (MNB4) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804464C100
Encompass_Preferred
Clone: MNB4. Isotype: IgG2a. Immunogen: Recombinant human PTX3 (pentraxin 3). Species reactivity: Human, Rat. Specificity: Recognizes the N-terminus of PTX3. Applications: ELISA, Flow Cytometry, IHC (FS), IP, WB. Formulation: Liquid. In PBS containing 0.1% sodium azide.
Catalog No. 50-200-9916
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.