Learn More
Clontech Labs 3P PUC 19 DNA 25 UGPUC 19 DNA 2
SDP
Supplier:  Clontech Labs 3P 3219
Encompass
Genomic DNA; pUC 19; 25ug; 0.5OD; 2,686bp; 250-1000u/mL
Catalog No. 50-443-743
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.