Learn More
AdipoGen Puerarin (250 mg)
SDP
Supplier:  AdipoGen CDXP0250M250
Encompass_Preferred
Chemical. CAS: 3681-99-0. Formula: C21H20O9. MW: 416.38. Isolated from plant source. Puerarin is a natural isoflavone isolated from plants of the genus Pueraria used in traditional Chinese herbal medicine. Daidzein and glucuronides are the main metabolites of puerarin. It has been widely used in the treatment of cardiovascular and cerebrovascular diseases, diabetes and diabetic complications, osteonecrosis, Parkinson's disease, Alzheimer's disease, endometriosis, and cancer. The beneficial effects of puerarin on the various medicinal purposes may be due to its wide spectrum of pharmacological properties such as vasodilation, cardioprotection, neuroprotection, antioxidant, anticancer, anti-inflammation, alleviating pain, promoting bone formation, inhibiting alcohol intake and attenuating insulin resistance. Due to the poor water-solubility and liposolubility, the applications of puerarin are limited.
Catalog No. 50-205-4596
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.