Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uniform Technology Pullover Hood with Darlex Around Face Opening Trioct, White

Catalog No. 17779028
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-779-028 Small
17-779-029 Medium
17-779-030 Large
17-779-031 2X-Large
17-779-032 3X-Large
17-779-034 X-Large
17-779-037 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 17-779-028 Supplier Uniform Technology Supplier No. CHCIG709WHS
This item is not returnable. View return policy

Uniform Technology Pullover Hood with integrated tricot face mask, 2-ply 929 sheer ear vents, and horizontal metal snaps in the back.

  • Pullover Hood
  • With darlex around face opening, integrated tricot face mask with pleats, 2-ply 929 sheer ear vents, 2-ply tricot vertical and horizontal metal snaps in back
  • Ideal for General CE Spaces/Applications (Non-ESD sensitive Medical Device, Electronics & Aerospace, Food Processing/Handling or Packaging, Cultivation Industry, University Labs, Imaging/Film coating Manufactures, General Medical/Dental Facilities)

Specifications

Size Small
Material Polyester, Carbon, Tricot
Color White
Disposable Reusable
Latex Free Yes
Seam Type Bound
Sterility Non-sterile
Length (Metric) 46.99 cm
Length (Metric) Facepiece 46.99 cm
For Use With (Application) General Cleanroom Spaces
Resistance Anti-static
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.