Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pullulan, MP Biomedicals™

Catalog No. MP215394405 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215394405 5 g
MP215394401 1 g
MP215394425 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP215394405 Supplier MP Biomedicals, Inc Supplier No. 0215394405
Add to Cart
Add to Cart

Produced from starch by cultivating a black yeast. Pullulan, MP Biomedicals is an edible plastic with excellent adhesiveness and film formability. It is a natural neutral polysaccharfied used in studies assessing bone tissue engineering and investigating

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9057-02-7
Molecular Formula (C18H30O15)AHO
InChI Key GFZFEWWPMNSVBS-WVZDODFGSA-N
Synonym Pullulan from Aureobasidium pullulans
PubChem CID 131636581
IUPAC Name (2R,4S,5S)-2-[(3S,4R,6R)-6-[(3S,4R,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-methoxyoxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(methoxymethyl)oxane-3,4,5-triol
SMILES COCC1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)OC)CO)CO)O)O)O

Specifications

Quantity 5 g
Solubility Information Dissolves easily in cold and hot water (50mg/mL; 1/20: Clear).
Formula Weight 200,000 (Lit.)
Physical Form Powder
Recommended Storage Store at Room Temperature (15–30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.