Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pullulan, Spectrum™ Chemical
SDP

Catalog No. 18605952 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-952 100 g Poly Bottle
18-605-951 25 g Poly Bottle
18-605-953 500 g Poly Pail
18-605-954 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-605-952 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P1830100GM
Add to Cart
Add to Cart

P1830, 9057-02-7, (C36H60O30)n

Spectrum™ Chemical Pullulan , a polysaccharide polymer, is used in the manufacture of edible films used in oral hygiene products. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 9057-02-7
Molecular Formula (C18H30O15)AHO
SMILES *.O-*.[H][C@]1(-*)O[C@H](CO)[C@@H](O[C@@]2([H])O[C@H](CO)[C@@H](O[C@@]3([H])O[C@H](CO-*)[C@@H](O)[C@H](O)[C@H]3O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 100 g
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.