Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pumice Stone, MilliporeSigma™

Catalog No. MPX19803 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2.5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MPX19803 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MPX19803 Supplier MilliporeSigma Supplier No. PX19803
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1332-09-8
Molecular Formula Al2O5Si
Molecular Weight (g/mol) 162.04
MDL Number MFCD00011232 MFCD00217788 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370732 MFCD07370733
InChI Key HNPSIPDUKPIQMN-UHFFFAOYSA-N
PubChem CID 76973482
IUPAC Name oxo[(oxoalumanyl)oxy]alumane; silanedione
SMILES O=[Si]=O.O=[Al]O[Al]=O

Specifications

Physical Form Powder
Quantity 2.5 kg
Chemical Name or Material Pumice Stone, Complex silicates of aluminum, potassium, and sodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.