Learn More
Julabo USA Inc Each, pump nozzle insulation for M16x1 fittings
SDP
Supplier:  Julabo USA Inc 8970751
Encompass_Preferred
Bath Accessories, Julabo, Pump nozzle insulation set for use with circulators having M16x1 female pump inlet/outlet connections. Install on pump connections to reduce ice build up / heat loss.
Catalog No. 50-753-4166
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.