Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

InterMetro Pump Sanitizer Stand

Catalog No. 11927003 Shop All Metro Products
Change view
Click to view available options
Mounting:
Freestanding
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
11-927-003 Freestanding
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-927-003 Supplier InterMetro Supplier No. HSSPA

InterMetro Pump Sanitizer includes mounting plates for both 1 gallon and 2 liter sanitizer pump dispensers.

Base of sanitizer sits 37 in. from the floor. Sanitizer not included.

Specifications

Color Black
Depth (English) 40 in.
Width (English) 10.5 in.
Height (English) 40 in.
Material Powder-coated Steel
Mounting Freestanding
Includes Mounting plates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.