Learn More
ILEX LIFE SCIENCES LLC Biocolor Purple-Jelley Hyaluronan Assay, Standard Size Kit (100 assays)
SDP
Supplier:  ILEX LIFE SCIENCES LLC H1000
Encompass_Preferred
The Biocolor Purple-Jelley Hyaluronan Assay is a quantitative colorimetric dye-binding assay used to measure the quantity of hyaluronic acid (HA) in mammalian tissue samples. It is a dye-based method with a detection limit of 0.2 μg. The dye used is ‘Stains-all’. The assay manual includes methods for the user to remove tissue protein and sulfated glycosaminoglycans (sGAG) before measuring the isolated hyaluronic acid. Also included in the manual is an outline guide to the distribution of hyaluronic acid present in various animal tissues – heart, kidney, lung, leg muscle and skin. All values were obtained using this assay. In adult animals the hyaluronic acid percentage gradually increases during ageing as muscle and fat mass decrease. Sensitivity: 0.2 µg. Synonyms: hyaluronic acid, HA, Purple-Jelly Hyaluronan Assay
Catalog No. 50-211-9999
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.