Learn More
Novus Biologicals™ Puratrophin-1 Recombinant Protein Antigen

Description
Specifications
Specifications
| Gene ID (Entrez) | 25894 |
| Purification Method | >80% by SDS-PAGE and Coomassie blue staining |
| Common Name | Puratrophin-1 Recombinant Protein Antigen |
| Content And Storage | Store at −20°C. Avoid freeze-thaw cycles. |
| Formulation | PBS and 1M Urea, pH 7.4. |
| For Use With (Application) | Blocking/Neutralizing, Control |
| Gene Alias | ARHGEF44, DKFZP434I216, PH domain-containing family G member 4, pleckstrin homology domain containing, family G (with RhoGef domain) member 4, Pleckstrin homology domain-containing family G member 4, PRTPHN1, puratrophin-1, Purkinje cell atrophy associate |
| Gene Symbol | PLEKHG4 |
| Label Type | Unlabeled |
| Product Type | Recombinant Protein Antigen |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.