Learn More
Biolegend Purified anti-EGF
Supplier:  Biolegend 679502
Encompass_Preferred
Purified anti-EGF [M1401E03]; Isotype: Mouse IgG1; Reactivity: Human; Apps: IP; Size: 100 ug
Catalog No. 50-208-7401