Learn More
Biolegend Purified anti-Insulin Degrading Enzyme (IDE)
Supplier:  Biolegend 921204
Encompass_Preferred
Purified anti-Insulin Degrading Enzyme (IDE) [9B12.225]; Isotype: Mouse IgG1, κ; Reactivity: Human, Rat, Hamster, Monkey; Apps: WB; Size: 100 ug
Catalog No. 50-207-5243
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.