Learn More
Biolegend Purified anti-mouse CD244.2 (2B4 B6 Alloantigen)
Supplier:  Biolegend 133501
Encompass_Preferred
Purified anti-mouse CD244.2 (2B4 B6 Alloantigen) [m2B4 (B6)458.1]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC, IP, IHC, ELISA; Size: 50 ug
Catalog No. 50-164-683
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.