Learn More
Biolegend Purified anti-mouse CD41
Supplier:  Biolegend 133901
Encompass_Preferred
Purified anti-mouse CD41 [MWReg30]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC, FA; Size: 50 ug
Catalog No. 50-164-701
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.