Learn More
Biolegend Purified anti-rat CD106
Supplier:  Biolegend 200402
Encompass_Preferred
Purified anti-rat CD106 [MR106]; Isotype: Mouse IgG1, κ; Reactivity: Rat; Apps: FC, IHC-F; Size: 500 ug
Catalog No. 50-165-304
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.