Learn More
Biolegend Purified anti-S100B
Supplier:  Biolegend 676604
Encompass_Preferred
Purified anti-S100B [11C12E12]; Isotype: Mouse IgG1; Reactivity: Human, Mouse, Rat; Apps: WB; Size: 100 ug
Catalog No. 50-208-7360