Learn More
Biolegend Purified anti-Tubulin β-3 (TUBB3)
Supplier:  Biolegend 802001
Encompass_Preferred
Purified anti-Tubulin β-3 (TUBB3) [Poly18020]; Isotype: Rabbit Polyclonal IgG; Reactivity: Human, Mouse, Rat; Apps: IHC-P, WB, ICC; Size: 100 uL
Catalog No. 50-102-9522
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.