Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIOTIX™ Pure uTIP™ Filter Sterile Universal Pipette Tips
GREENER_CHOICE

Catalog No. 12111141 Shop All OHAUS Products
Change view
Click to view available options
Volume (Metric):
2 μL
10 μL
20 μL
100 μL
200 μL
300 μL
1000 μL
1250 μL
Tip Style:
Ergonomic, Extended Length, Fine Point, Finely Tapered, Non-beveled Point, Eppendorf Style
Ergonomic, Extended Length, Finely Tapered, Flextop, Non-beveled Point
Ergonomic, Fine Point, Finely Tapered, Flextop, Non-beveled Point
Ergonomic, Finely Tapered, Fine Point, Flextop, Non-beveled Point
Ergonomic, Flextop, Fine Point, Finely Tapered, Non-beveled Point
Extended Length, Fine Point, Finely Tapered, Ergonomic
Fine Point, Finely Tapered, Ergonomic
Fine Point, Finely Tapered, Flextop, Ergonomic
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Tip Style
12-111-141 10 μL Ergonomic, Extended Length, Fine Point, Finely Tapered, Non-beveled Point, Eppendorf Style
12-111-171 2 μL Ergonomic, Finely Tapered, Fine Point, Flextop, Non-beveled Point
12-111-170 10 μL Fine Point, Finely Tapered, Flextop, Ergonomic
12-111-169 10 μL Extended Length, Fine Point, Finely Tapered, Ergonomic
12-111-168 20 μL Fine Point, Finely Tapered, Ergonomic
12-111-167 100 μL Ergonomic, Flextop, Fine Point, Finely Tapered, Non-beveled Point
12-111-166 200 μL Ergonomic, Fine Point, Finely Tapered, Flextop, Non-beveled Point
12-111-165 300 μL Ergonomic, Fine Point, Finely Tapered, Flextop, Non-beveled Point
12-111-164 1000 μL Ergonomic, Fine Point, Finely Tapered, Flextop, Non-beveled Point
12-111-163 1250 μL Ergonomic, Extended Length, Finely Tapered, Flextop, Non-beveled Point
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 12-111-141 Supplier BIOTIX™ Supplier No. 63305451
Add to Cart
Add to Cart
This item is not returnable. View return policy

Pure uTIP™ Filter Pipette Tips for Universal Pipettes, Standard Designed with Flexfit™ to lower necessary insertion and ejection force by ∼70% to make secure seal. Manufactured with Pure resin that is virgin polypropylene.

Biotix™ Pure uTIP™ Filter Pipette Tips for Universal Pipettes, Standard Designed with Flexfit™ to lower necessary insertion and ejection force by about 70% to make secure seal. Biotix™ uTIP™ Filter Pipette Tips for Universal Pipettes are manufactured with Pure resin that is virgin polypropylene.

  • Fits all standard pipettes
  • FlexFit technology features flexible proximal end to ensure secure seal with 66 to 80% less insertion force than competitor tips
  • Pure resin made of virgin polypropylene
  • Blade technology lowers chance of hanging droplet formation by optimizing surface area at distal end of tip
  • Certified free of RNase, DNase, Pyrogens, and PCR Inhibitors
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 10 μL
Filtered Filtered
Sterility Sterile
Color Clear
Rack Color Red
Autoclavable Not Autoclavable
Length (English) 1.80 in.
Clean Claims Sterile, Filtered, PCR Inhibitor Free, Endotoxin/Pyrogen Free, Nucleic Acid Free, RNase/DNase Free
Format Hinged Rack Sterile
Tip Style Ergonomic, Extended Length, Fine Point, Finely Tapered, Non-beveled Point, Eppendorf Style
Packaging Racked
Quantity 10 Racks of 96/Pk., 5 Pk./Cs.
Product Type Pipette Tips
Color Code Red
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line uTip
Material Polypropylene
Fit for Purpose Sterile and PCR Ready
Show More Show Less

This product contains no safety hazards

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.